Learn More
PRKAB2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human PRKAB2 peptide using ARM Technology.
Supplier: Abnova Corporation H00005565K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and noncatalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit may be a positive regulator of AMPK activity. It is highly expressed in skeletal muscle and thus may have tissue-specific roles. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| PRKAB2 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| PRKAB2 | |
| PRKAB2 | |
| A synthetic peptide of human PRKAB2 | |
| RUO | |
| 5565 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| protein kinase, AMP-activated, beta 2 noncatalytic subunit | |
| PRKAB2 | |
| MGC61468 | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.