Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PRKACA, Clone: 1C4, Abnova™

Catalog No. 89300219 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-219 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-219 Supplier Abnova Corporation Supplier No. H00005566M01A

Mouse Monoclonal Antibody

Specifications

Antigen PRKACA
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant PRKACA.
Formulation In ascites fluid
Gene Accession No. BC039846
Gene Alias MGC102831/MGC48865/PKACA
Gene Symbols PRKACA
Host Species Mouse
Immunogen PRKACA (AAH39846, 1 a.a. ∼ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5566
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Ascites
Isotype IgG Mix κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.