Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein kinase, interferon-inducible double stranded RNA dependent activator, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016949 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-949 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-949 Supplier Abnova Corporation Supplier No. H00008575B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PRKRA protein.

This gene encodes a protein kinase activated by double-stranded RNA which mediates the effects of interferon in response to viral infection. Mutations in this gene have been associated with dystonia. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Sequence: MSQSRHRAEAPPLEREDSGTFSLGKMITAKPGKTPIQVLHEYGMKTKNIPVYECERSDVQIHVPTFTFRVTVGDITCTGEGTSKKLAKHRAAEAAINILKANASICFAVPDPLMPDPSKQPKNQLNPIGSLQELAIHHGWRLPEYTLSQEGGPAHKREYTTICRLESFMETGKGASKKQAKRNAAEKFLAKFSNISPENHISLTNVVGHSLGCTWHSLRNSPGEKINLLKRSLLSIPNTDYIQLLSEIAKEQGFNITYLDIDELSANGQYQCLAELSTSPITVCHGSGISCGNAQSDAAHNALQYLKIIAERK

Specifications

Antigen protein kinase, interferon-inducible double stranded RNA dependent activator
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PRKRA protein.
Formulation PBS with no preservative; pH 7.4
Gene PRKRA
Gene Accession No. NM_003690
Gene Alias DYT16/HSD14/PACT/RAX
Gene Symbols PRKRA
Host Species Mouse
Immunogen PRKRA (NP_003681.1, 1 a.a. ∼ 313 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8575
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.