Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKR interacting protein 1 (IL11 inducible), Mouse, Clone: 4D11-3F11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant PRKRIP1.
Supplier: Abnova Corporation H00079706M05
Description
Sequence: MASPAASSVRPPRPKKEPQTLVIPKNAAEEQKLKLERLMKNPDKAVPIPEKMSEWAPRPPPEFVRDVMGSSAGAGSGEFHVYRHLRRREYQRQDYMDAMAEKQKLYAEFQKRLEKNKIAAEEQTAKRRKKRQKLKEKKLLAKKMKLEQKKQEGPGQPKEQGSSSSAEASGTEEEEEVPSFTMGRSpecifications
PRKR interacting protein 1 (IL11 inducible) | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC014298 | |
PRKRIP1 | |
PRKRIP1 (AAH14298, 1 a.a. ∼ 184 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
79706 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Immunofluorescence, Western Blot | |
4D11-3F11 | |
Mouse monoclonal antibody raised against a full length recombinant PRKRIP1. | |
PRKRIP1 | |
C114/FLJ13902/FLJ40957 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction