Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRL-2 Mouse anti-Human, Mouse, Clone: 42, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051583
Description
Anti-PRL-2 Antibody, clone 42 detects level of PRL-2 and has been published and validated for use in western blot, immunohistochemistry and immunoprecipitation.
Specifications
PRL-2 | |
Monoclonal | |
Unconjugated | |
NP_001158217.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
42 | |
Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
ptp4a2, HH13, HH7-2, HU-PP-1, OV-1, PRL-2, PRL2, PTP(CAAXII), PTP4A, PTPCAAX2, ptp-IV1a, ptp-IV1b | |
GST-tagged recombinant protein corresponding to mouse PRL-2. | |
100 μg | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction