Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protamine 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016889 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-889 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-889 Supplier Abnova Corporation Supplier No. H00005619B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PRM1 protein.

Sequence: MARYRCCRSQSRSRYYRQRQRSRRRRRRSCQTRRRAMRCCRPRYRPRCRRH

Specifications

Antigen protamine 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PRM1 protein.
Formulation PBS with no preservative; pH 7.4
Gene PRM1
Gene Accession No. BC003673
Gene Alias P1
Gene Symbols PRM1
Host Species Mouse
Immunogen PRM1 (AAH03673, 1 a.a. ∼ 51 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5619
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.