Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT3 NT Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB3341MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3341MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3341MI Supplier MilliporeSigma Supplier No. AB3341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect PRMT3 N-terminal with Anti-PRMT3 N-terminal Antibody (Rabbit Polyclonal Antibody), that has been shown to work in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT3 NT
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.09% sodium azide.
Gene Symbols PRMT3; HRMT1L3; EC 2.1.1.-
Host Species Rabbit
Immunogen Synthetic peptide from N-terminus of human PRMT3.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005788.2
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.