Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRMT5 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3349
Description
Anti-PRMT5 Antibody, C-terminus is a high quality Rabbit Polyclonal Antibody for the detection of PRMT5 and has been validated in ELISA and western blotting.
Specifications
PRMT5 | |
Polyclonal | |
Purified immunoglobulin in PBS containing 0.09% sodium azide. | |
Rabbit | |
100 μg | |
NM_001039619.1; NM_006109.3 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
ELISA, Western Blot | |
Unconjugated | |
PRMT5; SKB1; HRMT1L5; IBP72; SKB1Hs; JBP1; EC 2.1.1.125; EC 2.1.1.- | |
Synthetic peptide from C-terminus of human PRMT5. | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction