Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT5 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07405MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-405-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-405-MI Supplier MilliporeSigma Supplier No. 07405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PRMT5 Antibody is a Rabbit Polyclonal Antibody for detection of PRMT5 also known as 72 kDa ICln-binding protein, HMT1 hnRNP methyltransferase-like 5, Jak-binding protein 1, SKB1 homolog and has been validated in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit IgG in buffer containing 70% storage buffer (0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide) and 30% glycerol. Store at −20°C.
Gene Symbols PRMT5; HRMT1L5; IBP72; SKB1Hs; JBP1; SKB1
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 60-79 (KNRPGPQTRSDLLLSGRDWN) of human protein arginine methyltransferase 5, SKB1 homolog. The immunizing sequence is identical in mouse.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_006109
Target Species Human, Mouse
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.