Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pro-Insulin Cptide Mouse anti-Human, Clone: CP-01, MilliporeSigma™

Catalog No. 051109MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-110-9MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-110-9MI Supplier MilliporeSigma Supplier No. 051109
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Pro-Insulin C-Peptide Antibody, clone C-PEP-01 is validated for use in immunohistochemistry (paraffin) for the detection of Pro-Insulin C-Peptide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pro-Insulin C-Peptide
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C-PEP-01
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1 in phosphate buffered saline, pH7.4, and 15mM sodium azide.
Gene Accession No. P01308
Gene Symbols ILPR, IRDN, OTTHUMP00000011161 , OTTHUMP00000011162, OTTHUMP00000196038, insulin, proinsulin
Host Species Mouse
Immunogen Synthetic C-peptide of human pro-insulin conjugated to bovine serum albumin.
Purification Method Sequential precipitation with caprylic acid and ammonium sulfate.
Quantity 1 mg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.