Learn More
Procollagen Type III PIIINP Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB764P
Description
Anti-Procollagen Type III PIIINP Antibody detects level of Procollagen Type III PIIINP and has been published and validated for use in ELISA, immunohistochemistry, RIA.
Specifications
Procollagen Type III PIIINP | |
Polyclonal | |
Liquid in 0.01M Phosphate, 0.09M NaCl, pH 7.2 | |
Immunoaffinity purified | |
Primary | |
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. |
ELISA, Immunohistochemistry, Radioimmune Assays (RIA) | |
Unconjugated | |
Rabbit | |
100 μg | |
Human | |