Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein C receptor, endothelial (EPCR), Mouse, Clone: M3, Abnova™

Catalog No. 89001184 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-184 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-184 Supplier Abnova Corporation Supplier No. H00010544M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PROCR.

The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy. [provided by RefSeq

Sequence: MLTTLLPILLLSGWAFCSQDASDGLQRLHMLQISYFRDPYHVWYQGNASLGGHLTHVLEGPDTNTTIIQLQPLQEPESWARTQSGLQSYLLQFHGLVRLVHQERTLAFPLTIRCFLGCELPPEGSRAHVFFEVAVNGSSFVSFRPERALWQADTQVTSGVVTFTLQQLNAYNRTRYELREFLEDTCVQYVQKHISAENTKGSQTSRSYTSLVLGVLVGGFIIAGVAVGIFLCTGGRRC

Specifications

Antigen protein C receptor, endothelial (EPCR)
Applications ELISA, Western Blot
Classification Monoclonal
Clone M3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant PROCR.
Formulation PBS with no preservative; pH 7.4
Gene PROCR
Gene Accession No. BC014451
Gene Alias CCCA/CCD41/CD201/EPCR/MGC23024/bA42O4.2
Gene Symbols PROCR
Host Species Mouse
Immunogen PROCR (AAH14451.1, 1 a.a. ∼ 238 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10544
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.