Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prodynorphin Guinea Pig anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5519MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5519MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5519MI Supplier MilliporeSigma Supplier No. AB5519
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Anti-Prodynorphin Antibody is an antibody against Prodynorphin for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prodynorphin
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified Guinea pig antibody in PBS containing 0.05% sodium azide.
Gene Symbols PDYN; MGC26418; preprodynorphin; leumorphin; beta-neoendorphin-dynorphin; rimorphin; prodynorphin; PENKB; Preprodynorphin; leu-enkephalin
Host Species Guinea Pig
Immunogen Synthetic peptide from rat ProDynorphin.
Purification Method Affinity Purified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_024411.2
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.