Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progerin Mouse anti-Human, Clone: 13A4, MilliporeSigma™

Catalog No. 051231MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-123-1MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-123-1MI Supplier MilliporeSigma Supplier No. 051231
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Progerin Antibody, clone 13A4 is validated for use in western blot, immunocytochemistry, ELISA, immunoprecipitation for the detection of Progerin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Progerin
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 13A4
Conjugate Unconjugated
Formulation Protein G purified monoclonal antibody from ascites. Supplied at 1mg/mL in 0.1MTris-Glycine (pH7.4), 150mM NaCl, 0.05% sodium azide.
Gene Accession No. NP_005563
Gene Symbols LMNA
Host Species Mouse
Immunogen KLH-conjugated synthetic peptide corresponding to the amino acids 604-611 of progerin (lamin A/C).
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.