Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

prokineticin 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89018863 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-863 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-863 Supplier Abnova Corporation Supplier No. H00060675A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PROK2.

This gene encodes a protein expressed in the suprachiasmatic nucleus (SCN) circadian clock that may function as the output component of the circadian clock. The secreted form of the encoded protein may also serve as a chemoattractant for neuronal precursor cells in the olfactory bulb. Proteins from other vertebrates which are similar to this gene product were isolated based on homology to snake venom and secretions from frog skin, and have been shown to have diverse functions. Mutations in this gene are associated with Kallmann syndrome 4. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: LTPRAGDAAVITGACDKDSQCGGGMCCAVSIWVKSIRICTPMGKLGDSCHPLTRKVPFFGRRMHHTCPCLPGLACLRTSFNRFICLAQK

Specifications

Antigen prokineticin 2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PROK2.
Formulation 50% glycerol
Gene PROK2
Gene Accession No. NM_021935
Gene Alias BV8/KAL4/MIT1/PK2
Gene Symbols PROK2
Host Species Mouse
Immunogen PROK2 (NP_068754, 20 a.a. ∼ 108 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 60675
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.