Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Prolyl 4-hydroxylase β, Clone: 6-9H6,

Catalog No. MAB2073MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2073MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2073MI Supplier MilliporeSigma™ Supplier No. MAB2073

Specifications

Antigen Prolyl 4-hydroxylase β
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6-9H6
Conjugate Unconjugated
Formulation Purified immunoglobulin
Gene Accession No. NM_000917.2; NM_001017962.1
Gene Symbols P4HA1, P4HA, EC 1.14.11.2
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.