Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCSK2 Rabbit anti-Human, Non-human primate, Rat, Polyclonal, Invitrogen™

Catalog No. PA184119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA184119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA184119 Supplier Thermo Scientific Supplier No. PA184119

Rabbit Polyclonal Antibody

Rabbit anti Human proprotein convertase 2 antibody recognizes the peptidase enzyme designated proprotein convertase 2, also known as PC2, NEC2 and neuroendocrine convertase 2.

Proprotein convertase 2, also known as PC2, NEC2 and neuroendocrine convertase 2, is a peptidase enzyme involved in the processing of hormone (and other protein) precursors and is localized in secretory granules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCSK2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene PCSK2
Gene Accession No. P16519, P28841
Gene Alias NEC2, PC2, SPC2
Gene Symbols PCSK2
Host Species Rabbit
Immunogen Proprietary Immunogen.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25121, 5126
Target Species Human, Non-human Primate, Rat
Content And Storage 4°C or -20°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.