Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostasin Mouse anti-Human, Unlabeled, Clone: 2, BD

Catalog No. BDB612173 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612173 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612173 Supplier BD Biosciences Supplier No. 612173
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prostasin is a serine protease expressed as both GPI-anchored and secreted forms. Prostasin displays trypsin-like enzymatic activity by hydrolyzing substrates, such as D-Pro-Phe-Arg-AMC. This activity is inhibited by aprotinin, antipain, leupeptin, and benzamidine. The highest levels of prostasin protein are found in semen and the prostate, but many other tissues express prostasin at low levels. The sequence of prostasin is similar to other proteases, such as prostase, testisin, plasma kallikrein, and acrosin. In seminal fluid, prostasin can be found in complex with prostasin-binding protein, which can inhibit prostasin enzymatic activity. Prostasin mRNA is found in normal prostate epithelial cells and non-invasive prostate cancer cells, but is not expressed in invasive prostate cancer cell lines. Expression of prostasin in the invasive prostate cancer cell lines, DU-145 and PC-3, reduces the invasiveness of the cells in vitro. Thus, prostasin is a serine protease that may have roles in normal prostate function and in suppression of tumor cell invasion.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Prostasin
Applications Western Blot
Classification Monoclonal
Clone 2
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Prostasin aa. 108-228
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.