Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prosurfactant Protein B Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3430
Description
Detect Prosurfactant Protein B using this Anti-Prosurfactant Protein B Antibody, reacts with both C-terminus and N-terminus validated for use in ELISA, immunohistochemistry and western blotting.
Specifications
Prosurfactant Protein B | |
Polyclonal | |
Rabbit serum. Liquid. Contains no preservative. | |
Rabbit | |
100 μL | |
NM_000542.2; NM_198843.1 | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
SFTPB; SP-B; SFTP3; Phe; PSP-B; SFTB3; SMDP1 | |
Full length recombinant peptide (381 a.a.) of the human SP-B (pre)proprotein. | |
Primary | |
Human, Mouse | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction