Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Prosurfactant Protein C (proSP-C) Rabbit anti-Human, Mouse, Rat, Polyclonal

Catalog No. AB3786MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3786MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3786MI Supplier MilliporeSigma™ Supplier No. AB3786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Prosurfactant Protein C (proSP-C) Antibody detects level of Prosurfactant Protein C (proSP-C) and has been published and validated for use in ELISA, Immunohistochemistry (immunohistochemistry), Immunohistochemistry - Paraffin (immunohistochemistry-P) and Western Blotting (WB).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prosurfactant Protein C (proSP-C)
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Polyclonal rabbit serum. Contains no preservative.
Gene Symbols SFTPC; SP-C; SFTP2; SMDP2; Val.; SP5; PSP-C
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-32 from the N-terminal of the human SP-C proprotein coupled to KLH
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003018.3
Target Species Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.