Learn More
Anti-Protein A Chicken Polyclonal Antibody HRP Conjugate , TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Specifications
| Antigen | Protein A Chicken |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Host Species | Chicken |
| Immunogen | Protein A (Staphylococcus aureus-derived) |
| Quantity | 1 Vial |
| Primary or Secondary | Primary |
| Content And Storage | Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. |
| Isotype | IgY |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.