Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein-arginine deiminase 4 Mouse anti-Human, Clone: 10F1, MilliporeSigma™

Catalog No. MABE254MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE254MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE254MI Supplier MilliporeSigma Supplier No. MABE254
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-Protein-arginine deiminase 4 Antibody, clone 10F1 (mouse monoclonal antibody) validated in western blot, immunoprecipitation to detect PAD-4 also known as HL-60 PAD, Protein-arginine deiminase type-4, Peptidylarginine deiminase IV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein-arginine deiminase 4
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10F1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_036519
Gene Symbols PADI4, PADI5, PDI5
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Protein-arginine deminiase 4.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.