Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Protein Disulfide Isomerase/P4HB, Polyclonal, Alexa Fluor 488, R&D Systems™

Catalog No. IC7986G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC7986G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC7986G Supplier R&D Systems Supplier No. IC7986G
This item is not returnable. View return policy

Host Species: Goat
Isotype: IgG
Species Reactivity: Hu
Immunogen: Chinese hamster ovary cell line CHO-derived recombinant human P4HB, Asp18-Lys505

Applications:

Immunocytochemistry

Specifications

Antigen Protein Disulfide Isomerase/P4HB
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Immunocytochemistry 1:10 dilution
Gene Accession No. P07237
Gene Alias Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDI, PDIA1, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHB, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, Thbp, thyroid hormone-binding protein p55
Gene Symbols P4HB
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human P4HB, Asp18-Lys505, Accession # P07237
Purification Method Affinity Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 5034
Test Specificity Designed to visualize the expression of P4HB by fluorescence microscopy for staining cells and tissues. Conjugated P4HB antibodies are ideal for immunocytochemistry colocalization studies in the lumen of the endoplasmic reticulum. The unconjugated antibody detects human P4HB in direct ELISAs and Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.