Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Protein Disulfide Isomerase, Polyclonal

Catalog No. 53922950UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
53-922-950UL 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-922-950UL Supplier MilliporeSigma™ Supplier No. 53922950UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Protein Disulfide Isomerase Clone: in Bovine, Canine, Guinea Pig, Hamster, Human, Monkey, Mouse, Porcine, Rat, Sheep, Xenopus samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation

PDI is an abundant component of the ER that catalyzes disulfide bond formation in the biosynthesis of various secretory and cell surface proteins. It is a multi-functional polypeptide identical to theβ-subunit of prolyl 4-hydroxylase and a cellular thyroid hormone-binding protein. It has two thioredoxin-like regions, each con-taining a Cys-Gly-His-Cys sequence that may represent two independently acting catalytic sites of PDI activity. The C-terminus of PDI has a KDEL sequence, which acts as retention signal for proteins of the ER lumen. PDI exists as a homodimer of∽107kDa. Prolyl-4-hydroxylase, anα2β2 tetramer, catalyzes the formation of 4-hydroxyproline in collagen by the hydroxylation of proline residues in peptide linkages. The PDI/β-subunit of prolyl-4-hydroxylase is required for its retention in the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein Disulfide Isomerase
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, 50% glycerol.
Host Species Rabbit
Immunogen bovine liver Protein Disulfide Isomerase
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽55kDa (apparent MW) protein disulfide isomerase protein.
Target Species Bovine, Canine, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep, Xenopus species
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.