Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Gene Product 9.5 Guinea Pig anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5898MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5898MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5898MI Supplier MilliporeSigma Supplier No. AB5898
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Anti-Protein Gene Product 9.5 Antibody detects level of Protein Gene Product 9.5 and has been published and validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein Gene Product 9.5
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid. Contains 0.05% sodium azide.
Gene Symbols UCHL1; PARK5; UCH-L1; Uch-L1; PGP9.5; EC 3.4.19.12; EC 6.-.-.-
Host Species Guinea Pig
Immunogen A 17 amino acid peptide corresponding to amino acids 175-191 from soluble cytoplasmic human PGP9.5.
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004181.3
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.