Learn More
Protein Gene Product 9.5 Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1761I
Description
Detect PGP9.5 using this rabbit polyclonal antibody, Anti-Protein Gene Product 9.5 Antibody validated for use in western blotting, immunohistochemistry (Paraffin), Immunofluorescence and immunocytochemistry.
Specifications
| Protein Gene Product 9.5 | |
| Polyclonal | |
| Purified rabbit polyclonal in buffer containing PBS with 0.1% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| UCHL1 | |
| KLH-conjugated linear peptide corresponding to human Protein Gene Product 9.5. | |
| 100 μg | |
| NP_004172 | |
| 2-8°C for 12 months |