Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Phosphatase 2 C α/Β Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071223MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-122-3MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-122-3MI Supplier MilliporeSigma Supplier No. 071223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Protein Phosphatase 2 C α/β using this Anti-Protein Phosphatase 2 C α/β Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein Phosphatase 2 C α/β
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin in phosphate buffered saline with 0.08% sodium azide.
Gene Symbols PPM2C; PDPC; PDP1; FLJ32517; PDH; PDP; MGC119646; EC 3.1.3.43
Host Species Rabbit
Immunogen Purified peptide conjugated to KLH.
Purification Method Protein Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) Q9P0J1
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.