Learn More
Proteinase-Activated Receptor 2 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Proteinase-Activated Receptor 2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Peptide-affinity purified immunoglobulin. Liquid in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide |
| Gene Symbols | F2RL1; PAR-2; PAR2; GPR11 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to a sequence within the N-terminal extracellular domain of human PAR2. The immunogen has 70% homology with mouse and 60% homology with rat. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.