Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prothrombin Mouse anti-Human, Clone: GMA-095, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05879
Description
This Anti-Prothrombin Antibody, clone GMA-095 is validated for use in ELISA, western blotting for the detection of Prothrombin.
Specifications
| Prothrombin | |
| Monoclonal | |
| Unconjugated | |
| F2, prothrombin, PT | |
| Purified human Prothrombin | |
| 100 μg | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| GMA-095 | |
| NM_000506 | |
| Mouse | |
| DEAE purified | |
| Primary | |
| −20°C for up to 24 months. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction