Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protocadherin-17 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP197757
Description
Protocadherin-17 Polyclonal specifically detects Protocadherin-17 in Mouse samples. It is validated for Western Blot, Peptide ELISA.Specifications
| Protocadherin-17 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| PCDH68protocadherin-68, PCH68Protocadherin-68, protocadherin 17, protocadherin 68, protocadherin-17 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27253 | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| NP_001035519.1 | |
| PCDH17 | |
| Peptide with sequence C-RVFADVHSRASRDS corresponding to internal region (near C-Terminus) according to NP_001035519.1. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Mouse. Expected from sequence similarity: Human, Rat, Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction