Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prox1 Mouse anti-Chicken, Human, Mouse, Rat, Lizard, Clone: 5G10, MilliporeSigma™

Catalog No. MAB5652MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5652MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5652MI Supplier MilliporeSigma Supplier No. MAB5652
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Prox1 Antibody, clone 5G10 is validated for use in western blot, immunohistochemistry for the detection of Prox1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prox1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5G10
Conjugate Unconjugated
Formulation Liquid in PBS with 0.1% sodium azide.
Gene Accession No. NM_002763.3
Gene Symbols PROX1
Host Species Mouse
Immunogen Recombinant human Prox1 protein.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Chicken, Human, Lizard, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.