Learn More
protease, serine, 21 (testisin), Mouse, Clone: 2E10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PRSS21.
Supplier: Abnova Corporation H00010942M01
Description
This gene encodes a cell-surface anchored serine protease, which is a member of the trypsin family of serine proteases. It is predicted to be active on peptide linkages involving the carboxyl group of lysine or arginine. The protein localizes to the cytoplasm and the plasma membrane of premeiotic testicular germ cells and it may be involved in progression of testicular tumors of germ cell origin. Alternative splicing of this gene results in three transcript variants encoding three different isoforms. [provided by RefSeq
Sequence: VSNIYLSPRYLGNSPYDIALVKLSAPVTYTKHIQPICLQASTFEFENRTDCWVTGWGYIKEDEALPSPHTLQEVQVAIINNSMCNHLFLKYSFRKDIFGDSpecifications
protease, serine, 21 (testisin) | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_006799 | |
PRSS21 | |
PRSS21 (NP_006790, 121 a.a. ∼ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
10942 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Immunoprecipitation | |
2E10 | |
Mouse monoclonal antibody raised against a partial recombinant PRSS21. | |
PRSS21 | |
ESP-1/ESP1/TEST1/TESTISIN | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.