Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

proteinase 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018394 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-394 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-394 Supplier Abnova Corporation Supplier No. H00005657D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PRTN3 protein.

Sequence: MAHRPPSPALASVLLALLLSGAARAAEIVGGHEAQPHSRPYMASLQMRGNPGSHFCGGTLIHPSFVLTAAHCLRDIPQRLVNVVLGAHNVRTQEPTQQHFSVAQVFLNNYDAENKLNDILLIQLSSPANLSASVATVQLPQQDQPVPHGTQCLAMGWGRVGAHDPPAQVLQELNVTVVTFFCRPHNICTFVPRRKAGICFGDSGGPLICDGIIQGIDSFVIWGCATRLFPDFFTRVALYVDWIRSTLRRVEAKGRP

Specifications

Antigen proteinase 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PRTN3 protein.
Formulation PBS with no preservative; pH 7.4
Gene PRTN3
Gene Accession No. BC096184
Gene Alias ACPA/AGP7/C-ANCA/MBT/P29/PR-3
Gene Symbols PRTN3
Host Species Rabbit
Immunogen PRTN3 (AAH96184.1, 1 a.a. ∼ 256 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 5657
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.