Learn More
phosphoserine aminotransferase 1, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq
Specifications
Specifications
| Antigen | phosphoserine aminotransferase 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant PSAT1. |
| Formulation | 50% glycerol |
| Gene | PSAT1 |
| Gene Accession No. | NM_058179 |
| Gene Alias | EPIP/MGC1460/PSA/PSAT |
| Gene Symbols | PSAT1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.