Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSD-95 Mouse, Unlabeled, Clone: 16, BD

Catalog No. BDB610496 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610496 150 μg
BDB610495 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610496 Supplier BD Biosciences Supplier No. 610496
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PostSynaptic Density (PSD) refers to a dense region of submembranous cytoskeleton found most prominently in postsynaptic membranes of the CNS. Possible functions for the PSD include regulation and aggregation of receptors,structural stabilization of the synaptic junction, and transduction of signals from membrane receptors. Some of the proteins associated with the PSD are fodrin, tubulin, calmodulin, CaM Kinase II, PSD-95, and PSD-93. PSD-95 (SAP90) is a protein that interacts with the NMDA receptor NMDAR2B, neuronal NOS(nNOS or bNOS), and other proteins. PSD-95 contains one SH3 domain in its carboxy-terminal domain, as well as three conserved repeat regions called GLGF or PDZ domains. nNOS, which is concentrated in synaptic junctions, also contains a PZD domain. PSD-95 and nNOS interact via their respective PZD domains, which may mediate the binding of nNOS to skeletal muscle syntrophin.

This antibody is routinely tested by western blot analysis. Other applications were tested in BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD-95
Applications Western Blot
Classification Monoclonal
Clone 16
Concentration 250μg/mL
Conjugate Unconjugated
Description PostSynaptic Density-95; SAP90
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.