Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PSD-95 Clone: 7E3-1B8,

Catalog No. CP35100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
CP35100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CP35100UL Supplier MilliporeSigma™ Supplier No. CP35100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PSD-95 Clone: 7E3-1B8 in Rat samples, and it is validated for Immunofluorescence, Immunoprecipitation

Effective synaptic transmission requires the precise distribution and spatial localization of neurotransmitter receptors and ion channels on the postsynaptic nerve terminal. An understanding of synaptic development is derived largely from studies of the vertebrate skeletal neuromuscular junction. At glutamatergic synapses, the membrane associated cytoskeletal protein PSD-95 (Post Synaptic Density 95kDa) is believed to promote receptor clustering via interactions with the NR2 subunit of the NMDA receptor and the Shaker-type potassium channel. These interactions are likely mediated by multiple repeating PDZ domains (90 amino acid repeats). Two additional proteins, homer and GRIP, have recently been shown to interact with the metabotropic and AMPA receptors, respectively. It is likely that these proteins serve major functions in the molecular scaffolding and signal transduction process, which occurs at the synaptic junction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSD-95
Applications Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 7E3-1B8
Conjugate Unconjugated
Formulation In PBS.
Host Species Mouse
Immunogen recombinant, rat PSD95
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽95kDa PSD95 protein.
Target Species Rat
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.