Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSD-95 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300294
Description
PSD-95 Polyclonal specifically detects PSD-95 in Mouse, Rat samples. It is validated for Western Blot.Specifications
PSD-95 | |
Polyclonal | |
Western Blot 1:1000 | |
P31016 | |
DLG4 | |
Synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH. Accession # P31016 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for ∼95k PSD-95 protein. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15 | |
Rabbit | |
95 kDa | |
0.1 mL | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
1742 | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction