Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSD95 Mouse anti-Rat, Mouse, Human, Clone: K28/43, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN68
Description
Detect PSD95 using this Anti-PSD95 Antibody, clone K28/43 validated for use in immunohistochemistry, western blotting and immunocytochemistry.
Specifications
PSD95 | |
Monoclonal | |
Unconjugated | |
NP_001356 | |
Mouse | |
Protein G Purified | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
K28/43 | |
Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
PSD-95, PSD95, SAP-90, SAP90 | |
Recombinant protein corresponding to human PSD95. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction