Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA Mouse anti-Human, Clone: 3/A12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC291
Description
Anti-PSMA Antibody, clone 3/A12 is a highly specific mouse monoclonal antibody, that targets PSMA and has been tested in Flow Cytometry, western blotting, Immunofluorescence, Activity Assay and Inhibition.
Specifications
PSMA | |
Monoclonal | |
Unconjugated | |
NP_001014986 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Functional Assay, Flow Cytometry, Immunofluorescence, Inhibition Assays, Western Blot | |
3/A12 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
FOLH1, FOLH, NAALAD1, PSM, PSMA | |
Recombinant protein corresponding to human PSMA. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction