Learn More
proteasome (prosome, macropain) subunit, alpha type, 8 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | proteasome (prosome, macropain) subunit, alpha type, 8 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant PSMA8. |
| Formulation | 50% glycerol |
| Gene | PSMA8 |
| Gene Accession No. | NM_144662 |
| Gene Alias | MGC26605/PSMA7L |
| Gene Symbols | PSMA8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.