Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

proteasome (prosome, macropain) 26S subunit, ATPase, 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018399 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-399 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-399 Supplier Abnova Corporation Supplier No. H00005702D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PSMC3 protein.

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 nonidentical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a nonlysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes one of the ATPase subunits, a member of the triple-A family of ATPases that have chaperone-like activity. This subunit may compete with PSMC2 for binding to the HIV tat protein to regulate the interaction between the viral protein and the transcription complex. A pseudogene has been identified on chromosome 9. [provided by RefSeq

Sequence: MNLLPNIESPVTRQEKMATVWDEAEQDGIGEEVLKMSTEEIIQRTRLLDSEIKIMKSEVLRVTHELQAMKDKIKENSEKIKVNKTLPYLVSNVIELLDVDPNDQEEDGANIDLDSQRKGKCAVIKTSTRQTYFLPVIGLVDAEKLKPGDLVGVNKDSYLILETLPTEYDSRVKAMEVDERPTEQYSDIGGLDKQIQELVEAIVLPMNHKEKFENLGIQPPKGVLMYGPPGTGKTLLARACAAQTKATFLKLAGPQLVQMFIGDGAKLVRDAFALAKEKAPSIIFIDELDAIGTKRFDSEKAGDREVQRTMLELLNQLDGFQPNTQVKVIAATNRVDILDPALLRSGRLDRKIEFPMPNEEARARIMQIHSRKMNVSPDVNYEELARCTDDFNGAQCKAVCVEAGMIALRRGATELTHEDYMEGILEVQAKKKANLQYYA

Specifications

Antigen proteasome (prosome, macropain) 26S subunit, ATPase, 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PSMC3 protein.
Formulation PBS with no preservative; pH 7.4
Gene PSMC3
Gene Accession No. NM_002804.3
Gene Alias MGC8487/TBP1
Gene Symbols PSMC3
Host Species Rabbit
Immunogen PSMC3 (NP_002795.2, 1 a.a. ∼ 439 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 5702
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.