Learn More
PSMD13, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human PSMD13 peptide using ARM Technology.
Supplier: Abnova Corporation H00005719K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 nonidentical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a nonlysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator. Two transcripts encoding different isoforms have been described. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
PSMD13 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
PSMD13 | |
PSMD13 | |
A synthetic peptide of human PSMD13 | |
RUO | |
5719 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
proteasome (prosome, macropain) 26S subunit, non-ATPase, 13 | |
PSMD13 | |
HSPC027, Rpn9, S11, p40.5 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.