Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

proteasome (prosome, macropain) activator subunit 2 (PA28 beta), Mouse, Clone: 3F11-1A3, Abnova™

Catalog No. 89000914 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-914 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-914 Supplier Abnova Corporation Supplier No. H00005721M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PSME2.

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 nonidentical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a nonlysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. The immunoproteasome contains an alternate regulator, referred to as the 11S regulator or PA28, that replaces the 19S regulator. Three subunits (alpha, beta and gamma) of the 11S regulator have been identified. This gene encodes the beta subunit of the 11S regulator, one of the two 11S subunits that is induced by gamma-interferon. Three beta and three alpha subunits combine to form a heterohexameric ring. Six pseudogenes have been identified on chromosomes 4, 5, 8, 10 and 13. [provided by RefSeq

Sequence: MAKPCGVRLSGEARKQVEVFRQNLFQEAEEFLYRFLPQKIIYLNQLLQEDSLNVADLTSLRAPLDIPIPDPPPKDDEMETDKQEKKEVPKCGFLPGNEKVLSLLALVKPEVWTLKEKCILVITWIQHLIPKIEDGNDFGVAIQEKVLERVNAVKTKVEAFQTTISKYFSERGDAVAKASKETHVMDYRALVHERDEAAYGELRAMVLDLRAFYAELYHIISSNLEKIVNPKGEEKPSMY

Specifications

Antigen proteasome (prosome, macropain) activator subunit 2 (PA28 beta)
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3F11-1A3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant PSME2.
Formulation PBS with no preservative; pH 7.4
Gene PSME2
Gene Accession No. BC019885
Gene Alias PA28B/PA28beta/REGbeta
Gene Symbols PSME2
Host Species Mouse
Immunogen PSME2 (AAH19885, 1 a.a. ∼ 239 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5721
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.