Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Psme3/PA28-γ Mouse, Unlabeled, Clone: 47, BD

Catalog No. BDB611180 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611180 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611180 Supplier BD Biosciences Supplier No. 611180

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

The 20S proteasome is the major constituent of the proteasomal complex that mediates degradation events, including the generation of antigenic peptides that interact with MHC class I. The 20S proteasome is a cylindrical shaped complex composed of four layers of rings, each with seven subunits. The inner rings contain α-subunits, while the outer rings contain β-catalytic subunits. The constituents of the PA28 activator complex are additional subunits with specialized roles in class I-mediated antigen presentation. PA28 is a ring-shaped structure with alternating α- and β-subunits. This complex binds the α-rings of 20S and stimulates its activity. Expression of the PA28 α- and β-subunits is strongly induced by IFN-γ. The protein product of the Psme3 gene, also known as the Ki antigen, is highly homologous to the PA28 α- and β-subunits and has been designated the PA28 γ-subunit. This protein forms a homohexamer that binds the 20S proteasome and is thought to modulate proteasome activity. The PA28 α- and β-subunits are located in the cytoplasm and in the nucleus, while the γ-subunit is almost exclusively nuclear.

Host Species: Mouse
Clone: 47
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse Psme3/PA28-γ aa. 45-147
Formula Weight [Chemical]: 36kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Psme3/PA28-γ
Applications Western Blot
Classification Monoclonal
Clone 47
Concentration 250μg/mL
Conjugate Unconjugated
Description PA28-γ
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Psme3/PA28-γ aa. 45-147
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.