Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTEN Mouse, Alexa Fluor 647, Clone: A2B1, BD

Catalog No. BDB560003 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560003 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560003 Supplier BD Biosciences Supplier No. 560003
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cancer can develop when cells escape normal growth control mechanisms through mutations in proto-oncogenes or tumor suppressor genes. A characteristic of most oncogene and tumor suppressor gene products is that they are components of signal transduction pathways that are essential for maintaining cellular homeostasis. PTEN (phosphatase and tensin homolog), also known as MMAC1 (mutated in multiple advanced cancers 1), is a tumor suppressor gene that is mutated at high frequency in multiple tumor types. The protein encoded by PTEN is a phosphatase that preferentially dephosphorylates phosphoinositide substrates. It is believed that a mechanism by which PTEN mutations cause tumors is the loss of its negative control on the phosphoinositide 3-kinase signaling pathway that regulates cell growth and survival. PTEN also plays a role in the maintenance of hematopoietic stem cells.

The A2B1 monoclonal antibody recognizes PTEN, regardless of phosphorylation status.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PTEN
Applications Flow Cytometry
Classification Monoclonal
Clone A2B1
Conjugate Alexa Fluor 647
Description MMAC1, TEP1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human C-terminal PTEN Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.