Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTEN Mouse anti-Human, Mouse, Rat, Clone: 6H2.1, MilliporeSigma™

Catalog No. 04035MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-035-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-035-MI Supplier MilliporeSigma Supplier No. 04035
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PTEN Antibody, clone 6H2.1 detects level of PTEN and has been published and validated for use in western blot, immunoprecipitation, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTEN
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6H2.1
Conjugate Unconjugated
Formulation Protein G purified mouse IgG in storage buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_000305.3
Gene Symbols PTEN, MMAC1, TEP1
Host Species Mouse
Immunogen C-terminal 100 amino acids of PTEN
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.