Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTEN Mouse, Unlabeled, Clone: A2B1, BD

Catalog No. BDB559600 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559600 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559600 Supplier BD Biosciences Supplier No. 559600
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cancer can develop when cells escape normal growth control mechanisms through mutations in proto-oncogenes or tumor suppressor genes. A characteristic of most oncogene and tumor suppressor gene products is that they are components of signal transduction pathways that are essential for maintaining cellular homeostasis. PTEN (phosphatase and tensin homolog detected on chromosome ten/MMAC or TEP1 (mutated in multiple advanced cancers) is a tumor suppressor gene that has been identified as mutated in multiple tumor types. PTEN is a phosphatase that acts on proteins and lipid second messengers. The ability of PTEN to cause tumors is thought to result from the loss of its negative control on the phosphoinositide 3-kinase signaling pathway which regulates cell growth and survival. Using a mutant mouse model for PTEN, Suzuki et al. demonstrated that these mice had elevated levels of the protein kinase Akt in tumors associated with loss of heterozygosity of mPTEN. Human PTEN is a polypeptide of 403 amino acids based on the nucleotide sequence and is observed to migrate at ∽55kDa in SDS/PAGE. The antibody recognizes human and mouse PTEN. A peptide representing amino acids 387-400 at the C-terminus of human PTEN was used as the immunogen.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PTEN
Applications Western Blot
Classification Monoclonal
Clone A2B1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description MMAC1, TEP1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PTEN peptide aa. 387-400
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.