Learn More
Description
PTEN/MMAC1, a tumor suppressor, has been implicated in a large number of human tumors and is conserved from humans to worms. Characterization of PTEN protein showed that it is a phosphatase that acts on proteins and on 3-phosphorylated phosphoinositides, and can therefore modulate signal-transduction pathways that involve lipid second messengers. Recent results indicate that at least part of its role is to regulate the activity of the serine/threonine kinase AKT/PKB, and thus influence cell survival signaling.
Host Species: Mouse
Clone: 10P03; same as A5
Isotype: IgG1/κ
Cross Reactivity: Human and Mouse. Others-not known.
Epitope: aa 388-400
Immunogen: A synthetic peptide corresponding to aa 388-400 (ENEPFDEDQHTQI) from the C-terminus of human PTEN
Molecular Weight: 60kDa
Positive Control: A431, NIH-3T3 cells
Cellular Localization: Cytoplasmic
Immunofluorescence, Immunoprecipitation, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | PTEN/MMAC1 Ab-3 |
| Classification | Monoclonal |
| Clone | 10P03; same as A5 |
| Concentration | 250μg/100μL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Target Species | Human, Mouse |
| Isotype | IgG1 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.