Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTF1A Mouse, Unlabeled, Clone: S25-763, BD

Catalog No. BDB564745 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564745 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564745 Supplier BD Biosciences Supplier No. 564745
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The S25-763 monoclonal antibody specifically binds PTF1A (Pancreas specific transcription factor 1a). PTF1A is a transcription factor that contains a basic helix-loop-helix motif domain and, in combination with p75 and p64, forms the pancreas transcription factor complex. PTF1A plays a roles in pancreatic and cerebellar development. In early pancreatic development, pancreatic bud cells can either continue to pancreatic organogenesis or regress to a duodenal fate, and PTF1A plays a role in this differentiation. In late pancreatic development, the expression of PTF1A is restricted to acinar cells where it is involved in the maintenance of exocrine-specific gene expression. PTF1A expression is often evaluated to monitor pancreatic lineage differentiation from pluripotent stem cells. In addition, PTF1A is expressed during retinal development, and mutations in PTF1A cause cerebellar deficiencies.

This mouse anti-human PTF1A monoclonal antibody, clone S25-763, has been demonstrated to cross-react with rat and mouse pancreatic acinar cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PTF1A
Applications Flow Cytometry
Classification Monoclonal
Clone S25-763
Concentration 0.5mg/mL
Conjugate Unconjugated
Description PTF1α; Ptf1a; Ptf1α; bHLHa29; PTF1-p48
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PTF1A Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.