Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTH1R Mouse anti-Human, Clone: 4D2, MilliporeSigma™

Catalog No. MABN688MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN688MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN688MI Supplier MilliporeSigma Supplier No. MABN688
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PTH1R Antibody, clone 4D2 is a highly specific mouse monoclonal antibody that targets PTHR1 and has been tested in Immunocytochemistry, Immunohistochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTH1R
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4D2
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Gene Symbols PFE, PTHR, PTHR1, MGC138426, MGC138452, PTH1R
Host Species Mouse
Immunogen Purified recombinant fragment of human PTH1R expressed in E. Coli.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.